Learn More
Medchemexpress LLC Propiolactone | 57-57-8 | 99.36% | 72.06 | 25 MG
SDP
Catalog No. 5000346766
Encompass_Preferred
Propiolactone (β-propiolactone; 2-Oxetanone) is a viral chemical inactivator that causes the infectious inactivation of viruses. It has been used to inactivate SARS-CoV and as a bacteriostatic agent to formulate the BPL-inactivated influenza virus vaccine (Flu-BPL).
  • Causes infectious inactivation of viruses.
  • Used as a bacteriostatic agent to formulate BPL-inactivated influenza virus vaccine.
  • Can be used for vaccine purification.
  • Inactivated virus loses infectivity in Vero cells.
Catalog No. 50-003-46766 Supplier Medchemexpress LLC Supplier No. HY1079311G
Propiolactone (β-propiolactone; 2-Oxetanone) is a viral chemical inactivator that causes the infectious inactivation of viruses. It has been used to inactivate SARS-CoV and as a bacteriostatic agent to formulate the BPL-inactivated influenza virus vaccine (Flu-BPL).
  • Causes infectious inactivation of viruses.
  • Used as a bacteriostatic agent to formulate BPL-inactivated influenza virus vaccine.
  • Can be used for vaccine purification.
  • Inactivated virus loses infectivity in Vero cells.