Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC β-propiolactone | 57-57-8 | MFCD00005169 | 99.4% | 72.06 g·mol⁻¹ | C3H4O2 | 100 MG
SDP

Catalog No. 5000241943
Encompass_Preferred

Propiolactone (β-propiolactone) is a reactive four-membered lactone used as a chemical viral inactivator and research reagent. It is supplied for analytical and inactivation applications and is intended for research use only, not for human or clinical use.

  • High purity (99.36%).
  • Molecular weight 72.06 g·mol⁻¹.
  • Molecular formula C3H4O2.
  • Synonyms include β-propiolactone and 2-oxetanone.
  • Available as a liquid 100 MG and as a 10 mM solution (1 mL) in DMSO.
  • Storage: pure -20°C up to 3 years; in solvent -80°C for 6 months or -20°C for 1 month.
  • Intended for laboratory research; controlled-substance restrictions may apply in some territories.

Catalog No. 50-002-41943 Supplier Medchemexpress LLC Supplier No. HY107931100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Propiolactone (β-propiolactone) is a reactive four-membered lactone used as a chemical viral inactivator and research reagent. It is supplied for analytical and inactivation applications and is intended for research use only, not for human or clinical use.

  • High purity (99.36%).
  • Molecular weight 72.06 g·mol⁻¹.
  • Molecular formula C3H4O2.
  • Synonyms include β-propiolactone and 2-oxetanone.
  • Available as a liquid 100 MG and as a 10 mM solution (1 mL) in DMSO.
  • Storage: pure -20°C up to 3 years; in solvent -80°C for 6 months or -20°C for 1 month.
  • Intended for laboratory research; controlled-substance restrictions may apply in some territories.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.