Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Propionanilide, 98%, Thermo Scientific™

Catalog No. AAH3183214
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH3183214 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH3183214 Supplier Thermo Scientific Chemicals Supplier No. H3183214

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
The propionanilide derivatives are the first developed non-steroidal antiandrogens and include drugs such as flutamide, hydroxyflutamide, nilutamide and bicalutamide.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 620-71-3
Molecular Formula C9H11NO
Molecular Weight (g/mol) 149.193
MDL Number MFCD00092530
InChI Key ZTHRQJQJODGZHV-UHFFFAOYSA-N
Synonym propionanilide, n-phenylpropionamide, propanamide, n-phenyl, propionamide, n-phenyl, unii-uyp5zqi00t, uyp5zqi00t, methylacetanilid, n-phenyl-propionamide, ch3ch2conhph
PubChem CID 12107
IUPAC Name N-phenylpropanamide
SMILES CCC(=O)NC1=CC=CC=C1

Specifications

Melting Point 104°C to 107°C
Density 1.175
Boiling Point 222°C
Odor Odorless
Quantity 25 g
Solubility Information Insoluble in water.
Formula Weight 149.19
Percent Purity 98%
Chemical Name or Material Propionanilide
Hazard Category H302
Hazard Statement GHS H Statement
H302
Harmful if swallowed.
Precautionary Statement P264b-P270-P301+P312-P330-P501c
EINECSNumber 210-649-3
RTECSNumber UE4600000
TSCA Yes
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.