Learn More
Propionic acid, 99%
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Suitable for battery materials development.
Propionic acid is used as a preservative for animal feed and food for human consumption. It acts as an intermediate in the production of polymers like cellulose-acetate-propionate, artificial flavorings, agrochemicals and in bleaching agents. In addition, it is used in the synthesis of morphans.
Solubility
Miscible with water, alcohol, diethyl ether and chloroform.
Notes
Incompatible with strong oxidizing agents.
Specifications
Specifications
| Melting Point | -21°C |
| Density | 0.992 |
| Boiling Point | 140°C to 141°C |
| Flash Point | 51°C (123°F) |
| Odor | Unpleasant |
| Refractive Index | 1.3875 |
| Linear Formula | CH3CH2CO2H |
| Quantity | 1000 g |
| UN Number | UN3463 |
| Beilstein | 506071 |
| Show More |
Safety and Handling
| Hazard Category | H226-H290-H314-H335 |
| Hazard Statement | GHS H Statement H314-H226 Causes severe skin burns and eye damage. Flammable liquid and vapor. |
| Precautionary Statement | P210-P233-P234-P235-P240-P241-P242-P243-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P390-P501c |
| DOTInformation | Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: PROPIONIC ACID |
| EINECSNumber | 201-176-3 |
| RTECSNumber | UE5950000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.