Learn More
Cayman Chemical PropIonIc AcId-d5 25mg
SDP
Catalog No. 502916564
Encompass_Preferred
An internal standard for the quantification of propionic acid by GC- or LC-MS
Catalog No. 50-291-6564 Supplier Cayman Chemical Supplier No. 3915925 MG
An internal standard for the quantification of propionic acid by GC- or LC-MS