Learn More
Cayman Chemical PropIonIc AcId-d5 50mg
SDP
Catalog No. 502918352
Encompass_Preferred
An internal standard for the quantification of propionic acid by GC- or LC-MS
Catalog No. 50-291-8352 Supplier Cayman Chemical Supplier No. 3915950 MG
An internal standard for the quantification of propionic acid by GC- or LC-MS