Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Propionic Acid Hemicalcium Salt, MP Biomedicals™

Catalog No. ICN10498791 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
1 kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN10498791 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN10498791 Supplier MP Biomedicals, Inc Supplier No. 0210498791
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

An antifungal agent. Soluble in water (1g/3mL); slightly soluble in methanol, ethanol; practically insoluble in acetone, benzene

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4075-81-4
Molecular Formula C6H10CaO4
Molecular Weight (g/mol) 186.22
MDL Number MFCD00167354
InChI Key BCZXFFBUYPCTSJ-UHFFFAOYSA-L
Synonym calcium propionate, calcium dipropionate, calcium propanoate, mycoban, propanoic acid, calcium salt, bioban-c, caswell no. 151, propionic acid, calcium salt, unii-8ai80040kw, hsdb 907
PubChem CID 19999
ChEBI CHEBI:81716
IUPAC Name calcium dipropanoate
SMILES [Ca++].CCC([O-])=O.CCC([O-])=O

Specifications

Color White
Quantity 1 kg
Formula Weight 93.1
Physical Form Powder
Chemical Name or Material Propionic Acid Hemicalcium Salt
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.