Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Propionic Acid, NF, 99.5-100.5%, Spectrum™ Chemical
SDP

Catalog No. 18605886 Shop All Spectrum Chemical Mfg. Corp. Products
Change view
Click to view available options
Quantity:
500 mL
1 L
4 L
20 L
Packaging:
Amber Glass Bottle
Poly Drum
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-605-886 20 L Poly Drum
18-605-883 500 mL Amber Glass Bottle
18-605-882 1 L Amber Glass Bottle
18-605-884 4 L Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18-605-886 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. P146420LTBL

missing translation for 'validMainframeMsg'

P1464, 79-09-4, C3H6O2

Spectrum™ Chemical Propionic Acid, NF is used as a preservative in pharmaceutical formulations. Spectrum Chemical manufactured NF grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Specifications

CAS 79-09-4
Packaging Poly Drum
Quantity 20 L
Percent Purity 99.5 to 100.5%
Grade NF
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.