Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Propionic acid, sodium salt, 99.0-100.5%, ACROS Organics™

Click to view available options
Quantity:
25g
1kg
5kg
Packaging:
Plastic bottle
Plastic drum
137-40-6
C3H5NaO2
96.06
MFCD00002759
JXKPEJDQGNYQSM-UHFFFAOYSA-M
sodium propionate,sodium propanoate,propionic acid sodium salt,propanoic acid, sodium salt,napropion,ocuseptine,deketon,impedex,keenate,propiofar
2723816
sodium;propanoate
CCC(=O)[O-].[Na+]
137-40-6
C3H5NaO2
96.06
MFCD00002759
JXKPEJDQGNYQSM-UHFFFAOYSA-M
sodium propionate,sodium propanoate,propionic acid sodium salt,propanoic acid, sodium salt,napropion,ocuseptine,deketon,impedex,keenate,propiofar
2723816
sodium;propanoate
CCC(=O)[O-].[Na+]
Melting Point 285°C to 286°C
Flash Point >250°C
Color White
Packaging Plastic bottle
Infrared Spectrum Authentic
Quantity 25g
Water 1% max. (K.F.)
Assay Percent Range 99.0-100.5%
Linear Formula C2H5CO2Na
Beilstein 02, 234
Merck Index 15, 8798
Solubility Information Solubility in water: very soluble. Other solubilities: soluble in alcohol
Formula Weight 96.06
Physical Form Crystalline Powder
Percent Purity 99.0 to 100.5% (on dried substance)
Chemical Name or Material Propionic acid, sodium salt
Show More Show Less

GHS H Statement:
Causes skin irritation.
Harmful in contact with skin.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge