Learn More
Apexbio Technology LLC PROPIONYL-CARNITINE-D3-HCL-5MG
SDP
Catalog No. 5000570254
Encompass_Preferred
Propionyl carnitine-d3 HCl (CAS No 2692623-94-0) is a stable isotope-labeled compound used in biomedical research particularly in studies related to mitochondrial function and energy metabolism Originating from modifications of naturally occurring carnitine it is involved in the transport of fatty acids across mitochondrial membranes for beta-oxidation This compound targets metabolic pathways and is often assessed for its bioactivity in vitro across a range of concentrations from nanomolar to low micromolar Propionyl carnitine-d3 HCl is commonly utilized in cell models and animal studies to investigate metabolic disorders and potential therapeutic effects It is also applied in drug screening programs to evaluate the efficacy of new metabolic modulators Experimental concentrations typically depend on the specific research design
Catalog No. 50-005-70254 Supplier Apexbio Technology LLC Supplier No. SL10305
Propionyl carnitine-d3 HCl (CAS No 2692623-94-0) is a stable isotope-labeled compound used in biomedical research particularly in studies related to mitochondrial function and energy metabolism Originating from modifications of naturally occurring carnitine it is involved in the transport of fatty acids across mitochondrial membranes for beta-oxidation This compound targets metabolic pathways and is often assessed for its bioactivity in vitro across a range of concentrations from nanomolar to low micromolar Propionyl carnitine-d3 HCl is commonly utilized in cell models and animal studies to investigate metabolic disorders and potential therapeutic effects It is also applied in drug screening programs to evaluate the efficacy of new metabolic modulators Experimental concentrations typically depend on the specific research design