Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Propionyl chloride, 99.5%, phosgene free

Catalog No. AC458970250
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC458970250 25 g
AC458975000 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. AC458970250 Supplier Thermo Scientific Chemicals Supplier No. 458970250
Only null left
Add to Cart
This item is not returnable. View return policy

CAS: 79-03-8 | C3H5ClO | 92.52 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 79-03-8
Molecular Formula C3H5ClO
Molecular Weight (g/mol) 92.52
InChI Key RZWZRACFZGVKFM-UHFFFAOYSA-N
Synonym propionyl chloride, propionic acid chloride, propionic chloride, propionylchloride, unii-mb6vl5omb9, mb6vl5omb9, propanoylchloride, propionyl cloride, propanoic chloride, propionyl-chloride
PubChem CID 62324
IUPAC Name propanoyl chloride
SMILES CCC(=O)Cl

Specifications

Melting Point -94.0°C
Density 1.0590g/mL
Boiling Point 80.0°C
Flash Point 6°C
Infrared Spectrum Authentic
Assay Percent Range 99.4% min. (GC)
Linear Formula C2H5COCl
Quantity 25 g
Specific Gravity 1.059
Solubility Information Solubility in water: reacts.
Viscosity 0.48 mPa.s (20°C)
Formula Weight 92.52
Percent Purity 99.5%
Chemical Name or Material Propionyl chloride
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Harmful if swallowed.
Toxic if inhaled.
Causes severe skin burns and eye damage.
Reacts violently with water.
Highly flammable liquid and vapor.
Precautionary Statement GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Immediately call
EINECSNumber 201-170-

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.