Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Propionyl-DL-carnitine chloride | 18828-58-5 | 98.1% | 253.72 | C10H20ClNO4 | 100 MG
SDP

Catalog No. 5000212034
Encompass_Preferred

Propionyl-DL-carnitine chloride is a carnitine derivative used as a biochemical research reagent, notably in studies of inflammation and energy metabolism. It is provided as a white to off-white solid with high purity and defined storage recommendations to preserve stability.

  • High purity (98.1%).
  • White to off-white solid appearance.
  • Molecular weight 253.72 g/mol.
  • CAS number 18828-58-5.
  • Powder storage: -20°C for long-term stability.
  • Suitable for biochemical and inflammation research applications.
  • Available in multiple research-scale quantities including 100 mg.

Catalog No. 50-002-12034 Supplier Medchemexpress LLC Supplier No. HY101018100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Propionyl-DL-carnitine chloride is a carnitine derivative used as a biochemical research reagent, notably in studies of inflammation and energy metabolism. It is provided as a white to off-white solid with high purity and defined storage recommendations to preserve stability.

  • High purity (98.1%).
  • White to off-white solid appearance.
  • Molecular weight 253.72 g/mol.
  • CAS number 18828-58-5.
  • Powder storage: -20°C for long-term stability.
  • Suitable for biochemical and inflammation research applications.
  • Available in multiple research-scale quantities including 100 mg.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.