Learn More
Medchemexpress LLC Propionyl-DL-carnitine chloride | 18828-58-5 | 98.1% | 253.72 g/mol | C10H20ClNO4 | 1 ML
SDP
Catalog No. 5000212144
Encompass_Preferred
Propionyl-DL-carnitine chloride is a carnitine derivative provided for research use, available as a solid and as a ready-to-use 10 mM solution in DMSO. Product data list CAS 18828-58-5, formula C10H20ClNO4, molecular weight 253.72 g/mol, and purity 98.10%.
  • Supplied as powder and as a 10 mM, 1 mL solution in DMSO.
  • High purity: 98.1% as specified by the manufacturer.
  • Good solubility in methanol and DMSO; ultrasonic assistance recommended.
  • Storage stability documented for powder and in-solution conditions.
  • Intended for biochemical research, including inflammation studies.
Catalog No. 50-002-12144 Supplier Medchemexpress LLC Supplier No. HY10101810MM 1ML
Propionyl-DL-carnitine chloride is a carnitine derivative provided for research use, available as a solid and as a ready-to-use 10 mM solution in DMSO. Product data list CAS 18828-58-5, formula C10H20ClNO4, molecular weight 253.72 g/mol, and purity 98.10%.
  • Supplied as powder and as a 10 mM, 1 mL solution in DMSO.
  • High purity: 98.1% as specified by the manufacturer.
  • Good solubility in methanol and DMSO; ultrasonic assistance recommended.
  • Storage stability documented for powder and in-solution conditions.
  • Intended for biochemical research, including inflammation studies.