Learn More
Medchemexpress LLC Propionyl-DL-carnitine chloride | 18828-58-5 | 98.1% | 253.72 g/mol | C10H20ClNO4 | 25 MG
SDP
Catalog No. 5000211857
Encompass_Preferred
Propionyl-DL-carnitine chloride is a carnitine derivative used in biochemical and pharmacological research, particularly for studies of inflammation and oxidative stress. It is suitable for cellular and animal models investigating carnitine metabolism, endothelial function, and erythrocyte responses.
  • Used in inflammation and oxidative stress research
  • Protects erythrocytes from oxidative damage
  • Supports endothelial cell function and reduces reactive oxygen species
  • Modifies plasma and urine carnitine concentrations in vivo
  • High purity: 98.1%
  • White to off-white solid
  • Store powder at -20°C (3 years) or 4°C (2 years)
Catalog No. 50-002-11857 Supplier Medchemexpress LLC Supplier No. HY10101825MG
Propionyl-DL-carnitine chloride is a carnitine derivative used in biochemical and pharmacological research, particularly for studies of inflammation and oxidative stress. It is suitable for cellular and animal models investigating carnitine metabolism, endothelial function, and erythrocyte responses.
  • Used in inflammation and oxidative stress research
  • Protects erythrocytes from oxidative damage
  • Supports endothelial cell function and reduces reactive oxygen species
  • Modifies plasma and urine carnitine concentrations in vivo
  • High purity: 98.1%
  • White to off-white solid
  • Store powder at -20°C (3 years) or 4°C (2 years)