Learn More
Medchemexpress LLC Propionyl-DL-carnitine chloride | 18828-58-5 | 98.1% | 253.72 g/mol | C10H20ClNO4 | 50 MG
SDP
Catalog No. 5000212145
Encompass_Preferred
Propionyl-DL-carnitine chloride is a carnitine derivative supplied as a research reagent for biochemical and cellular studies. It is used in inflammation research and in studies of oxidative stress, cellular metabolism, and endothelial function.
  • High purity (98.1%) suitable for analytical and biological assays.
  • Protects beta-thalassaemic erythrocytes from oxidative stress.
  • Reduces formation of reactive oxygen species and supports endothelial function.
  • Increases production of 14CO2 from [1-14C]pyruvate in metabolic assays.
  • Affects plasma and urine total carnitine concentrations in vivo.
  • Available in small research quantities such as 50 mg.
Catalog No. 50-002-12145 Supplier Medchemexpress LLC Supplier No. HY10101850MG
Propionyl-DL-carnitine chloride is a carnitine derivative supplied as a research reagent for biochemical and cellular studies. It is used in inflammation research and in studies of oxidative stress, cellular metabolism, and endothelial function.
  • High purity (98.1%) suitable for analytical and biological assays.
  • Protects beta-thalassaemic erythrocytes from oxidative stress.
  • Reduces formation of reactive oxygen species and supports endothelial function.
  • Increases production of 14CO2 from [1-14C]pyruvate in metabolic assays.
  • Affects plasma and urine total carnitine concentrations in vivo.
  • Available in small research quantities such as 50 mg.