Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Propionyl-L-carnitine | 20064-19-1 | 95.0% | 10 MG
SDP

Catalog No. 5000234817
Encompass_Preferred

Propionyl-L-carnitine is an O-acyl L-carnitine derivative used in biochemical and pharmacological research to study energy metabolism and mitochondrial function. Supplied as a white to off-white solid with research-grade purity.

  • O-acyl L-carnitine derivative used to probe energy metabolism and mitochondrial function.
  • White to off-white solid suitable for analytical and in vitro studies.
  • Purity 95.0% as supplied for research use.
  • Molecular weight 217.26; formula C10H19NO4.
  • Store powder at -20°C or 4°C per product guidance.

Catalog No. 50-002-34817 Supplier Medchemexpress LLC Supplier No. HY12170510MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Propionyl-L-carnitine is an O-acyl L-carnitine derivative used in biochemical and pharmacological research to study energy metabolism and mitochondrial function. Supplied as a white to off-white solid with research-grade purity.

  • O-acyl L-carnitine derivative used to probe energy metabolism and mitochondrial function.
  • White to off-white solid suitable for analytical and in vitro studies.
  • Purity 95.0% as supplied for research use.
  • Molecular weight 217.26; formula C10H19NO4.
  • Store powder at -20°C or 4°C per product guidance.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.