Learn More
AdipoGen Propionyl-L-carnitine . hydrochloride (25 mg)
SDP
Supplier:  AdipoGen AGCR13595M025
Encompass_Preferred
Chemical. CAS: 119793-66-7. Formula: C10H19NO4 . HCl. MW: 217.3 . 36.5. Synthetic. Naturally occurring carnitine derivative formed by carnitine acetyltransferase during beta-oxidation of uneven chain fatty acids, with high affinity for muscular carnitine transferase. Increases cellular carnitine content, allowing free fatty acid transport into the mitochondria. Stimulates energy production in ischaemic muscles by increasing citric acid cycle flux and stimulating pyruvate dehydrogenase activity. Important for mitochondrial metabolism and energy regulation. Regulates the metabolism of both carbohydrates and lipids, leading to an increase of ATP generation. Selectively inhibits in vitro and ex vivo platelet-activating factor (PAF) synthesis from human neutrophils. Antioxidant. Shows free radical scavenging activity.
Catalog No. 50-205-3962