Learn More
Medchemexpress LLC Propionylcarnitine | 17298-37-2 | 98.0% | 217.26 | 1 MG
SDP
Catalog No. 5000436136
Encompass_Preferred
Propionylcarnitine is metabolized by carnitine acetyltransferase from propionyl-CoA. Increased propionylcarnitine is considered a biomarker of vitamin B12 deficiency. This product is intended for research use only.
  • Human endogenous metabolite
  • Protection of beta-thalassaemic erythrocytes from oxidative stress
  • Increases production of 14CO2 from [1-14C]pyruvate and increases the rate of formation of acetyl carnitine from pyruvate
  • Allows endothelial cells to maintain functionality and regulatory role on vessel activity for a longer time, and decreases the formation of oxygen reactive species due to xanthine oxidase activity on hypoxanthine
  • Affects plasma and urine total carnitine concentrations of mice (2 mM/kg; p.o. for 4 weeks)
Catalog No. 50-004-36136 Supplier Medchemexpress LLC Supplier No. HY1130921MG
Propionylcarnitine is metabolized by carnitine acetyltransferase from propionyl-CoA. Increased propionylcarnitine is considered a biomarker of vitamin B12 deficiency. This product is intended for research use only.
  • Human endogenous metabolite
  • Protection of beta-thalassaemic erythrocytes from oxidative stress
  • Increases production of 14CO2 from [1-14C]pyruvate and increases the rate of formation of acetyl carnitine from pyruvate
  • Allows endothelial cells to maintain functionality and regulatory role on vessel activity for a longer time, and decreases the formation of oxygen reactive species due to xanthine oxidase activity on hypoxanthine
  • Affects plasma and urine total carnitine concentrations of mice (2 mM/kg; p.o. for 4 weeks)