Learn More
Medchemexpress LLC Propionylcarnitine | 17298-37-2 | 98.0% | 217.26 | 10 MG
SDP
Catalog No. 5000436473
Encompass_Preferred
Propionylcarnitine is metabolized by carnitine acetyltransferase from propionyl-CoA. Increased propionylcarnitine is regarded as a biomarker of vitamin B12 deficiency.
  • Regarded as a biomarker of vitamin B12 deficiency.
  • Shows protection to beta-thalassaemic erythrocytes from oxidative stress.
  • Increases production of 14CO2 from [1-14C]pyruvate and increases the rate of formation of acetyl carnitine from pyruvate.
  • Allows endothelial cells to maintain their functionality and regulatory role on vessel activity for a longer time, and decreases the formation of oxygen reactive species due to xanthine oxidase activity on hypoxanthine formed by adenine nucleotide catabolism.
Catalog No. 50-004-36473 Supplier Medchemexpress LLC Supplier No. HY11309210MG
Propionylcarnitine is metabolized by carnitine acetyltransferase from propionyl-CoA. Increased propionylcarnitine is regarded as a biomarker of vitamin B12 deficiency.
  • Regarded as a biomarker of vitamin B12 deficiency.
  • Shows protection to beta-thalassaemic erythrocytes from oxidative stress.
  • Increases production of 14CO2 from [1-14C]pyruvate and increases the rate of formation of acetyl carnitine from pyruvate.
  • Allows endothelial cells to maintain their functionality and regulatory role on vessel activity for a longer time, and decreases the formation of oxygen reactive species due to xanthine oxidase activity on hypoxanthine formed by adenine nucleotide catabolism.