Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Propionylglycine | 21709-90-0 | MFCD09049674 | 98.0% | 131.13 | C5H9NO3 | 250 MG
SDP

Catalog No. 5000280473
Encompass_Preferred

Propionylglycine is a small N-acyl amino acid provided as a solid research reagent with documented purity, solubility, and storage recommendations suitable for biochemical studies.

  • High purity: 98.0%.
  • CAS number: 21709-90-0.
  • Chemical formula: C5H9NO3; molecular weight: 131.13.
  • Solubility: highly soluble in DMSO; soluble in recommended in vivo vehicle at reported concentrations.
  • Storage: solid at 4°C under inert atmosphere; stock solutions at -80°C (6 months) or -20°C (1 month).
  • Available as 250 mg solid and as a 10 mM solution in DMSO.

Catalog No. 50-002-80473 Supplier Medchemexpress LLC Supplier No. HYW587803250MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Propionylglycine is a small N-acyl amino acid provided as a solid research reagent with documented purity, solubility, and storage recommendations suitable for biochemical studies.

  • High purity: 98.0%.
  • CAS number: 21709-90-0.
  • Chemical formula: C5H9NO3; molecular weight: 131.13.
  • Solubility: highly soluble in DMSO; soluble in recommended in vivo vehicle at reported concentrations.
  • Storage: solid at 4°C under inert atmosphere; stock solutions at -80°C (6 months) or -20°C (1 month).
  • Available as 250 mg solid and as a 10 mM solution in DMSO.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.