Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Propionylpromazine-d6 hydrochloride | 1262770-67-1 | 99.8% | 382.98 | C20H19D6ClN2OS | 1 MG
SDP

Catalog No. 5000209086
Encompass_Preferred

Propionylpromazine-d6 hydrochloride is a deuterium-labeled hydrochloride salt of propionylpromazine used as a stable isotope standard and tracer in analytical workflows. The compound (C20H19D6ClN2OS) has a molecular weight of 382.98 and is suitable for use in LC-MS, GC-MS, and NMR quantitation.

  • Deuterium-labeled (d6) internal standard for quantitative mass spectrometry and NMR.
  • High purity suitable for analytical applications (99.8%).
  • Hydrochloride salt form with formula C20H19D6ClN2OS and MW 382.98.

Catalog No. 50-002-09086 Supplier Medchemexpress LLC Supplier No. HYW040146S1MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Propionylpromazine-d6 hydrochloride is a deuterium-labeled hydrochloride salt of propionylpromazine used as a stable isotope standard and tracer in analytical workflows. The compound (C20H19D6ClN2OS) has a molecular weight of 382.98 and is suitable for use in LC-MS, GC-MS, and NMR quantitation.

  • Deuterium-labeled (d6) internal standard for quantitative mass spectrometry and NMR.
  • High purity suitable for analytical applications (99.8%).
  • Hydrochloride salt form with formula C20H19D6ClN2OS and MW 382.98.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.