Learn More
Medchemexpress LLC Propionylpromazine-d6 hydrochloride | 1262770-67-1 | 99.8% | 382.98 g/mol | C20H19D6ClN2OS | 10 MG
SDP
Catalog No. 5000209233
Encompass_Preferred
Propionylpromazine-d6 hydrochloride is the deuterium-labeled (d6) isotopologue of propionylpromazine hydrochloride supplied as the hydrochloride salt. It is intended for use as a tracer or internal standard in quantitative analyses by NMR, GC-MS, or LC-MS, and is suitable for research involving dopamine receptor pharmacology.
  • Deuterium-labeled internal standard (d6).
  • Hydrochloride salt form for stability and solubility.
  • High purity suitable for quantitative analysis (99.8%).
  • Useful as tracer for NMR, GC-MS, and LC-MS methods.
  • Available in small analytical pack sizes for laboratory use.
Catalog No. 50-002-09233 Supplier Medchemexpress LLC Supplier No. HYW040146S10MG
Propionylpromazine-d6 hydrochloride is the deuterium-labeled (d6) isotopologue of propionylpromazine hydrochloride supplied as the hydrochloride salt. It is intended for use as a tracer or internal standard in quantitative analyses by NMR, GC-MS, or LC-MS, and is suitable for research involving dopamine receptor pharmacology.
  • Deuterium-labeled internal standard (d6).
  • Hydrochloride salt form for stability and solubility.
  • High purity suitable for quantitative analysis (99.8%).
  • Useful as tracer for NMR, GC-MS, and LC-MS methods.
  • Available in small analytical pack sizes for laboratory use.