Learn More
Medchemexpress LLC Propionylpromazine-d6 hydrochloride | 1262770-67-1 | 99.8% | 382.98 g/mol | C20H19D6ClN2OS | 5 MG
SDP
Catalog No. 5000209647
Encompass_Preferred
Propionylpromazine-d6 hydrochloride is the deuterium-labeled hydrochloride salt of propionylpromazine supplied as a high-purity research standard for analytical and pharmacological studies.
  • High isotopic purity suitable for mass spectrometry internal standards.
  • Provided as a hydrochloride salt for stability and solubility.
  • Available in small, controlled milligram quantities for analytical workflows.
  • Typical purity specification near 99.8% for accurate quantitation.
  • Store sealed, away from moisture and light; follow supplier storage guidance.
Catalog No. 50-002-09647 Supplier Medchemexpress LLC Supplier No. HYW040146S5MG
Propionylpromazine-d6 hydrochloride is the deuterium-labeled hydrochloride salt of propionylpromazine supplied as a high-purity research standard for analytical and pharmacological studies.
  • High isotopic purity suitable for mass spectrometry internal standards.
  • Provided as a hydrochloride salt for stability and solubility.
  • Available in small, controlled milligram quantities for analytical workflows.
  • Typical purity specification near 99.8% for accurate quantitation.
  • Store sealed, away from moisture and light; follow supplier storage guidance.