Learn More
Medchemexpress LLC HY-W040146 10mg Medchemexpress, Propionylpromazine (hydrochloride) CAS:7681-67-6 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYW04014610MG
Encompass_Preferred
Medchemexpress, HY-W040146 10mg Propionylpromazine (hydrochloride) CAS:7681-67-6 Propionylpromazine hydrochloride (Propiopromazine hydrochloride), a dopamine receptor D2 (DRD2) antagonist, can be used in the research of Parkinson disease. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-3465