Learn More
Cayman Chemical PropIophenon 10mg
SDP
Catalog No. 502943693
Encompass_Preferred
An analytical reference standard categorized as a precursor in the synthesis of cathinones; intended for research and forensic applications
Catalog No. 50-294-3693 Supplier Cayman Chemical Supplier No. 4190410 MG
An analytical reference standard categorized as a precursor in the synthesis of cathinones; intended for research and forensic applications