Learn More
Cayman Chemical Propiverine (hydrochloride), Purity: ≥98%, CAS: 54556-98-8, 100 mg
SDP
Catalog No. 502911631
Encompass_Preferred
A muscarinic acetylcholine receptor antagonist; binds to M1 and M2 mAChRs in rat bladder and submaxillary gland, respectively (Kis = 0.3 and 0.19 µM, respectively); inhibits carbachol-induced contraction of bladder strips isolated from patients with normal bladder activity and detrusor overactivity associated with benign prostatic hyperplasia (pA2s = 7.7 and 7.6, respectively)
Catalog No. 50-291-1631 Supplier Cayman Chemical Supplier No. 36049100 MG
A muscarinic acetylcholine receptor antagonist; binds to M1 and M2 mAChRs in rat bladder and submaxillary gland, respectively (Kis = 0.3 and 0.19 µM, respectively); inhibits carbachol-induced contraction of bladder strips isolated from patients with normal bladder activity and detrusor overactivity associated with benign prostatic hyperplasia (pA2s = 7.7 and 7.6, respectively)