Learn More
Electron Microscopy Sciences ProPlate™ MicroArray Slide Slide Module 1 Well, 16.5 x 70 mm I.D., Delrin Snap Clips
SDP
Catalog No. 5034317
Encompass_Preferred
ProPlate re-usable, multi-well chambers fit 25 x 75 mm glass microscope slide substrates. Chambers depth of 7.5 mm provides a generous surface to volume ratio to facilitate mixing and washing steps.
I.D.: 16.5 x 70 mm
Includes: Delrin Snap Clips
Catalog No. 50-343-17 Supplier Electron Microscopy Sciences Supplier No. 6348431
ProPlate re-usable, multi-well chambers fit 25 x 75 mm glass microscope slide substrates. Chambers depth of 7.5 mm provides a generous surface to volume ratio to facilitate mixing and washing steps.
I.D.: 16.5 x 70 mm
Includes: Delrin Snap Clips