Learn More
Electron Microscopy Sciences ProPlate™ MicroArray Slide Slide Module 1 Well, 16.5 x 70 mm I.D.
SDP
Catalog No. 5034285
Encompass_Preferred
ProPlate re-usable, multi-well chambers fit 25 x 75 mm glass microscope slide substrates. Chambers depth of 7.5 mm provides a generous surface to volume ratio to facilitate mixing and washing steps.
I.D.: 16.5 x 70 mm
Catalog No. 50-342-85 Supplier Electron Microscopy Sciences Supplier No. 6348421
ProPlate re-usable, multi-well chambers fit 25 x 75 mm glass microscope slide substrates. Chambers depth of 7.5 mm provides a generous surface to volume ratio to facilitate mixing and washing steps.
I.D.: 16.5 x 70 mm