Learn More
Electron Microscopy Sciences ProPlate™ MicroArray Slide Slide Module 16 Well, 6 mm I.D.
SDP
Catalog No. 5034320
Encompass_Preferred
ProPlate re-usable, multi-well chambers fit 25 x 75 mm glass microscope slide substrates. Chambers depth of 7.5 mm provides a generous surface to volume ratio to facilitate mixing and washing steps.
I.D.: 6 mm
Catalog No. 50-343-20 Supplier Electron Microscopy Sciences Supplier No. 6348426
ProPlate re-usable, multi-well chambers fit 25 x 75 mm glass microscope slide substrates. Chambers depth of 7.5 mm provides a generous surface to volume ratio to facilitate mixing and washing steps.
I.D.: 6 mm