Learn More
Electron Microscopy Sciences ProPlate™ MicroArray Slide Slide Module 16 Well, 7 x 7 mm I.D.
SDP
Catalog No. 5034321
Encompass_Preferred
ProPlate re-usable, multi-well chambers fit 25 x 75 mm glass microscope slide substrates. Chambers depth of 7.5 mm provides a generous surface to volume ratio to facilitate mixing and washing steps.
I.D.: 7 x 7 mm
Catalog No. 50-343-21 Supplier Electron Microscopy Sciences Supplier No. 6348427
ProPlate re-usable, multi-well chambers fit 25 x 75 mm glass microscope slide substrates. Chambers depth of 7.5 mm provides a generous surface to volume ratio to facilitate mixing and washing steps.
I.D.: 7 x 7 mm