Learn More
Electron Microscopy Sciences ProPlate™ MicroArray Slide Slide Module 3 Well, 18 x 20 mm I.D., Stainless Steel Clips
SDP
Catalog No. 5034331
Encompass_Preferred
ProPlate re-usable, multi-well chambers fit 25 x 75 mm glass microscope slides. Chambers depth of 7.5 mm. Stainless steel Spring Clips are used with laser fabricated glass slides that may have micro-fractures or chipped edges.
I.D.: 18 x 20 mm
Includes: Stainless Steel Clips
Catalog No. 50-343-31 Supplier Electron Microscopy Sciences Supplier No. 6348443
ProPlate re-usable, multi-well chambers fit 25 x 75 mm glass microscope slides. Chambers depth of 7.5 mm. Stainless steel Spring Clips are used with laser fabricated glass slides that may have micro-fractures or chipped edges.
I.D.: 18 x 20 mm
Includes: Stainless Steel Clips