Learn More
Electron Microscopy Sciences ProPlate™ MicroArray Slide Slide Module 64 Well, 3.5 x 3.5 mm I.D., Delrin Snap Clips
SDP
Catalog No. 5034319
Encompass_Preferred
ProPlate re-usable, multi-well chambers fit 25 x 75 mm glass microscope slides. Chambers depth of 7.5 mm. Delrin Snap Clips are easily applied and removed, allowing up to 4 chambered slides to be inserted into a tray.
I.D.: 3.5 x 3.5 mm
Includes: Delrin Snap Clips
Catalog No. 50-343-19 Supplier Electron Microscopy Sciences Supplier No. 6348438
ProPlate re-usable, multi-well chambers fit 25 x 75 mm glass microscope slides. Chambers depth of 7.5 mm. Delrin Snap Clips are easily applied and removed, allowing up to 4 chambered slides to be inserted into a tray.
I.D.: 3.5 x 3.5 mm
Includes: Delrin Snap Clips