Learn More
Electron Microscopy Sciences ProPlate™ MicroArray Slide Slide Module 8 Well, 7 x 16 mm I.D., Delrin Snap Clips
SDP
Catalog No. 5034325
Encompass_Preferred
ProPlate re-usable, multi-well chambers fit 25 x 75 mm glass microscope slides. Chambers depth of 7.5 mm. Delrin Snap Clips are easily applied and removed, allowing up to 4 chambered slides to be inserted into a tray.
I.D.: 7 x 16 mm
Includes: Delrin Snap Clips
Catalog No. 50-343-25 Supplier Electron Microscopy Sciences Supplier No. 6348435
ProPlate re-usable, multi-well chambers fit 25 x 75 mm glass microscope slides. Chambers depth of 7.5 mm. Delrin Snap Clips are easily applied and removed, allowing up to 4 chambered slides to be inserted into a tray.
I.D.: 7 x 16 mm
Includes: Delrin Snap Clips