Learn More
Electron Microscopy Sciences ProPlate™ MicroArray Slide Tray Set 8 Well, 7 x 16 mm I.D., Stainless Steel Clips
SDP
Catalog No. 5034295
Encompass_Preferred
ProPlate integrates microscope slide based array technology with automated microtiter plate processing. Up to 64 individual arrays can be assayed per slide. Four slides fit into a tray, producing a modular plate with a standard microtiter plate footprint and well spacing allowing automated robotic processing of up to 256 arrays.
Up to 64 leak-proof wells on any slide surface without the use of adhesive. Microtiter plate well spacing facilitates use with multichannel pipettes. 7 x 7 mm square well are ideal for high content arrays. 5 – 350 uL well volumes accommodate small reagent and large wash volumes.
Each ProPlate Tray Set includes: 4 Slide modules, 8 Snap clips, 1 Tray and Cover, 10 seal strips and one applicator. Tray conforms to SBS standards for footprint (81.7 mm width x 123.7 mm length) and well spacing.
I.D.: 7 x 16 mm
Includes: 8 Stainless Steel Spring Clips, Tray, Cover, 10 Seal Strips and 1 Seal Strip applicator
Catalog No. 50-342-95 Supplier Electron Microscopy Sciences Supplier No. 6348405
ProPlate integrates microscope slide based array technology with automated microtiter plate processing. Up to 64 individual arrays can be assayed per slide. Four slides fit into a tray, producing a modular plate with a standard microtiter plate footprint and well spacing allowing automated robotic processing of up to 256 arrays.
Up to 64 leak-proof wells on any slide surface without the use of adhesive. Microtiter plate well spacing facilitates use with multichannel pipettes. 7 x 7 mm square well are ideal for high content arrays. 5 – 350 uL well volumes accommodate small reagent and large wash volumes.
Each ProPlate Tray Set includes: 4 Slide modules, 8 Snap clips, 1 Tray and Cover, 10 seal strips and one applicator. Tray conforms to SBS standards for footprint (81.7 mm width x 123.7 mm length) and well spacing.
I.D.: 7 x 16 mm
Includes: 8 Stainless Steel Spring Clips, Tray, Cover, 10 Seal Strips and 1 Seal Strip applicator