Learn More
Electron Microscopy Sciences ProPlate™ MicroArray Slide Delrin Non Numbered Snap Clips
SDP
Catalog No. 5034312
Encompass_Preferred
ProPlate Delrin Snap Clips are easily applied and removed, allowing up to 4 chambered slides to be inserted into a tray.
Catalog No. 50-343-12 Supplier Electron Microscopy Sciences Supplier No. 6348481
ProPlate Delrin Snap Clips are easily applied and removed, allowing up to 4 chambered slides to be inserted into a tray.