Learn More
Grace Bio Labs Inc PROPLATE MULTI-WELL CHAMBER2PK
Catalog No. NC1625772
Encompass_Preferred
This item has a minimum qty of 2 per supplier requirements.
ProPlate 16 Well Slide Module, 7 mm x 7 mm x 7.6 mm Depth, Stainless Steel Spring Clips, Silicone gasket well size 7.5 mm x 7.5 mm 2 PACK
Catalog No. NC1625772 Supplier Grace Bio Labs Inc Supplier No. 244864
This item has a minimum qty of 2 per supplier requirements.
ProPlate 16 Well Slide Module, 7 mm x 7 mm x 7.6 mm Depth, Stainless Steel Spring Clips, Silicone gasket well size 7.5 mm x 7.5 mm 2 PACK