Learn More
Electron Microscopy Sciences ProPlate™ MicroArray Slide Stainless Steel Snap Clips
SDP
Catalog No. 5034313
Encompass_Preferred
ProPlate stainless steel Spring Clips are easily applied and removed and are preferred by customers using glass slides fabricated by means of a laser and which may have small micro-fractures from processing or chipped edges.
Catalog No. 50-343-13 Supplier Electron Microscopy Sciences Supplier No. 6348482
ProPlate stainless steel Spring Clips are easily applied and removed and are preferred by customers using glass slides fabricated by means of a laser and which may have small micro-fractures from processing or chipped edges.