Learn More
Electron Microscopy Sciences ProPlate™ MicroArray Slide Tray & Cover for Spring Clip Modules
SDP
Catalog No. 5034315
Encompass_Preferred
Organize up to 3 chambered slides into the footprint of a microtiter plate for use with automated liquid handling robots or plate washers.
Catalog No. 50-343-15 Supplier Electron Microscopy Sciences Supplier No. 6348484
Organize up to 3 chambered slides into the footprint of a microtiter plate for use with automated liquid handling robots or plate washers.