Learn More
Electron Microscopy Sciences ProPlate™ MicroArray Slide Tray & Cover for Snap Clip Modules
SDP
Catalog No. 5034314
Encompass_Preferred
Organize up to 4 chambered slides into the footprint of a microtiter plate for use with automated liquid handling robots or plate washers.
Catalog No. 50-343-14 Supplier Electron Microscopy Sciences Supplier No. 6348483
Organize up to 4 chambered slides into the footprint of a microtiter plate for use with automated liquid handling robots or plate washers.