Learn More
Med Vet International Propoflo Vet Use Intravenous Anesthetic inj 20mLmust Have Vet License On File
Catalog No. 502828554
Encompass_Preferred
VET LICENSE NEEDS TO BE PROVIDED IN 3-4 BUSINESS DAYS OR ORDER WILL BE CANCELLED
Propoflo is an intravenous anesthetic injection for use in dogs as follows: for induction of anesthesia, maintenance of general anesthesia for up to 20 minutes, and general anesthesia where maintenance is provided by inhalant anesthetics.
Catalog No. 50-282-8554 Supplier Med Vet International Supplier No. RXVPROPOFLO
VET LICENSE NEEDS TO BE PROVIDED IN 3-4 BUSINESS DAYS OR ORDER WILL BE CANCELLED
Propoflo is an intravenous anesthetic injection for use in dogs as follows: for induction of anesthesia, maintenance of general anesthesia for up to 20 minutes, and general anesthesia where maintenance is provided by inhalant anesthetics.