Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Propofol-d17 β-D-glucuronide | 1683581-05-6 | 98.0% | 371.50 | C18H9D17O7 | 10 MG
SDP

Catalog No. 5000229422
Encompass_Preferred

Propofol-d17 β-D-glucuronide is a deuterium-labeled analytical standard of propofol β-D-glucuronide intended as a tracer or internal standard for quantitative methods including NMR, GC-MS, and LC-MS. It is supplied as a high-purity solid with defined solubility and storage conditions to support reliable analytical workflows.

  • Deuterium-labeled internal standard for quantitative analysis.
  • High purity (98.0%) suitable for reference and calibration.
  • Supplied as a white to off-white solid powder.
  • Soluble in ethanol, DMF, and DMSO at ≥ 30 mg/mL.
  • Recommended storage: powder at -20°C; solutions at -80°C (6 months) or -20°C (1 month).
  • Available in small pack sizes appropriate for analytical use.

Catalog No. 50-002-29422 Supplier Medchemexpress LLC Supplier No. HY118774S10MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Propofol-d17 β-D-glucuronide is a deuterium-labeled analytical standard of propofol β-D-glucuronide intended as a tracer or internal standard for quantitative methods including NMR, GC-MS, and LC-MS. It is supplied as a high-purity solid with defined solubility and storage conditions to support reliable analytical workflows.

  • Deuterium-labeled internal standard for quantitative analysis.
  • High purity (98.0%) suitable for reference and calibration.
  • Supplied as a white to off-white solid powder.
  • Soluble in ethanol, DMF, and DMSO at ≥ 30 mg/mL.
  • Recommended storage: powder at -20°C; solutions at -80°C (6 months) or -20°C (1 month).
  • Available in small pack sizes appropriate for analytical use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.