Learn More
Medchemexpress LLC Propofol-d17 β-D-glucuronide | 1683581-05-6 | 98.0% | 371.50 g/mol | C18H9D17O7 | 1 MG
SDP
Catalog No. 5000226416
Encompass_Preferred
Propofol-d17 β-D-glucuronide is a deuterium-labeled internal standard of propofol β-D-glucuronide suitable for use as a tracer or quantitative reference in NMR, GC-MS, and LC-MS analyses. It is supplied as a high-purity isotopologue for metabolite quantitation and method development.
  • Deuterium-labeled propofol β-D-glucuronide for use as an internal standard
  • High purity: 98.0%
  • Available in small quantities including 1 mg
  • Soluble in ethanol, DMF, and DMSO at ≥ 30 mg/mL
  • Powder: store at -20°C; in solvent: -80°C (6 months) or -20°C (1 month)
  • Suitable for NMR, GC-MS, and LC-MS quantitative analysis
Catalog No. 50-002-26416 Supplier Medchemexpress LLC Supplier No. HY118774S1MG
Propofol-d17 β-D-glucuronide is a deuterium-labeled internal standard of propofol β-D-glucuronide suitable for use as a tracer or quantitative reference in NMR, GC-MS, and LC-MS analyses. It is supplied as a high-purity isotopologue for metabolite quantitation and method development.
  • Deuterium-labeled propofol β-D-glucuronide for use as an internal standard
  • High purity: 98.0%
  • Available in small quantities including 1 mg
  • Soluble in ethanol, DMF, and DMSO at ≥ 30 mg/mL
  • Powder: store at -20°C; in solvent: -80°C (6 months) or -20°C (1 month)
  • Suitable for NMR, GC-MS, and LC-MS quantitative analysis