Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Propofol-d17 β-D-glucuronide | 1683581-05-6 | 98.0% | 371.50 g/mol | C18H9D17O7 | 100 UG
SDP

Catalog No. 5000226292
Encompass_Preferred

Propofol-d17 β-D-glucuronide is a deuterium-labeled analogue of propofol β-D-glucuronide intended for use as a tracer and internal standard in quantitative analytical methods (NMR, GC-MS, LC-MS). It is provided as a white to off-white solid with high purity and characterized molecular properties, and is soluble in common organic solvents for analytical sample preparation.

  • Deuterium-labeled internal standard for quantitative analysis.
  • High purity: 98.0%.
  • Molecular weight 371.50 g/mol and formula C18H9D17O7.
  • Soluble in ethanol, DMF, and DMSO (≥30 mg/mL).
  • Available in microgram to milligram pack sizes for analytical use.
  • Suitable for use as a tracer in metabolic and pharmacokinetic studies.
  • Handle with care: use newly opened DMSO due to hygroscopicity.

Catalog No. 50-002-26292 Supplier Medchemexpress LLC Supplier No. HY118774S100UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Propofol-d17 β-D-glucuronide is a deuterium-labeled analogue of propofol β-D-glucuronide intended for use as a tracer and internal standard in quantitative analytical methods (NMR, GC-MS, LC-MS). It is provided as a white to off-white solid with high purity and characterized molecular properties, and is soluble in common organic solvents for analytical sample preparation.

  • Deuterium-labeled internal standard for quantitative analysis.
  • High purity: 98.0%.
  • Molecular weight 371.50 g/mol and formula C18H9D17O7.
  • Soluble in ethanol, DMF, and DMSO (≥30 mg/mL).
  • Available in microgram to milligram pack sizes for analytical use.
  • Suitable for use as a tracer in metabolic and pharmacokinetic studies.
  • Handle with care: use newly opened DMSO due to hygroscopicity.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.