Learn More
Cayman Chemical Propoxur-d3 10mg
SDP
Catalog No. 502918281
Encompass_Preferred
An internal standard for the quantification of propoxur by GC- or LC-MS
Catalog No. 50-291-8281 Supplier Cayman Chemical Supplier No. 3958210 MG
An internal standard for the quantification of propoxur by GC- or LC-MS