Learn More
Medchemexpress LLC Propoxycaine hydrochloride | 550-83-4 | 99.7% | 1 ML
SDP
Catalog No. 5000412808
Encompass_Preferred
Propoxycaine hydrochloride is an inhibitor of voltage-gated sodium channels, which prevents the ionic flux necessary for the initiation and conduction of impulses. Its application leads to a loss of sensation.
  • Inhibits voltage-gated sodium channels
  • Prevents ionic flux for impulse initiation and conduction
  • Leads to a loss of sensation
Catalog No. 50-004-12808 Supplier Medchemexpress LLC Supplier No. HYB124310MM/1ML
Propoxycaine hydrochloride is an inhibitor of voltage-gated sodium channels, which prevents the ionic flux necessary for the initiation and conduction of impulses. Its application leads to a loss of sensation.
  • Inhibits voltage-gated sodium channels
  • Prevents ionic flux for impulse initiation and conduction
  • Leads to a loss of sensation