Learn More
Medchemexpress LLC Propoxycaine hydrochloride | 550-83-4 | 99.7% | 100 MG
SDP
Catalog No. 5000411928
Encompass_Preferred
Propoxycaine hydrochloride inhibits voltage-gated sodium channels, and thereby inhibits the ionic flux required for the initiation and conduction of impulses. Propoxycaine hydrochloride application can lead to a loss of sensation.
  • Sodium channel inhibitor
  • Inhibits voltage-gated sodium channels
  • Inhibits the ionic flux required for the initiation and conduction of impulses
  • Can lead to a loss of sensation
Catalog No. 50-004-11928 Supplier Medchemexpress LLC Supplier No. HYB1243100MG
Propoxycaine hydrochloride inhibits voltage-gated sodium channels, and thereby inhibits the ionic flux required for the initiation and conduction of impulses. Propoxycaine hydrochloride application can lead to a loss of sensation.
  • Sodium channel inhibitor
  • Inhibits voltage-gated sodium channels
  • Inhibits the ionic flux required for the initiation and conduction of impulses
  • Can lead to a loss of sensation