Learn More
Medchemexpress LLC Propoxycaine hydrochloride | 550-83-4 | 99.7% | 5 MG
SDP
Supplier:  Medchemexpress LLC HYB12435MG
Encompass_Preferred
Propoxycaine hydrochloride inhibits voltage-gated sodium channels, thereby inhibiting the ionic flux required for the initiation and conduction of impulses. Its application can lead to a loss of sensation. This laboratory chemical is intended for research use only and should be handled by qualified and experienced scientists in appropriately equipped and authorized facilities.
  • Inhibits voltage-gated sodium channels
  • Inhibits ionic flux for impulse initiation and conduction
  • Leads to a loss of sensation
  • Stable under recommended storage conditions
  • Requires personal protective equipment for handling
Catalog No. 50-004-11872