Learn More
Medchemexpress LLC Propoxycaine hydrochloride | 550-83-4 | MFCD01660627 | 99.7% | 330.85 | C16H27ClN2O3 | 10 MG

Supplier: Medchemexpress LLC HYB124310MG
Propoxycaine hydrochloride is an ester-type local anesthetic and sodium-channel inhibitor supplied as a solid research reagent. It is used in laboratory studies requiring blockade of voltage-gated sodium channels and is provided with documentation for purity and storage.
- Inhibits voltage-gated sodium channels, reducing nerve impulse conduction.
- Supplied as a white to light brown solid suitable for in vitro use.
- Molecular formula C16H27ClN2O3 and molecular weight 330.85 g/mol.
- High purity (≈99.7-99.98%) with available certificate of analysis.
- Storage recommendations: 4°C sealed for solid; in solvent -80°C (6 months) or -20°C (1 month).
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.