Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Propoxyphenyl aildenafil | 1391053-82-9 | >98.0% | C24H34N6O4S | 10MG
SDP

Catalog No. 5000260542
Encompass_Preferred

Propoxyphenyl Aildenafil is a sildenafil analogue research compound used to study enhancement of sexual function and related pharmacology. It is a small-molecule PDE5 inhibitor analogue with molecular formula C24H34N6O4S and molecular weight 502.63 g/mol.

  • Sildenafil analogue for PDE5-related research.
  • Useful in preclinical studies of sexual function enhancement.
  • Molecular formula C24H34N6O4S.
  • Molecular weight 502.63 g/mol.
  • Available in multiple small-scale pack sizes for laboratory use.
  • Reported purity >98% suitable for research use.

Catalog No. 50-002-60542 Supplier Medchemexpress LLC Supplier No. HY13138310MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Propoxyphenyl Aildenafil is a sildenafil analogue research compound used to study enhancement of sexual function and related pharmacology. It is a small-molecule PDE5 inhibitor analogue with molecular formula C24H34N6O4S and molecular weight 502.63 g/mol.

  • Sildenafil analogue for PDE5-related research.
  • Useful in preclinical studies of sexual function enhancement.
  • Molecular formula C24H34N6O4S.
  • Molecular weight 502.63 g/mol.
  • Available in multiple small-scale pack sizes for laboratory use.
  • Reported purity >98% suitable for research use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.