Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MSC PROPPER Flight Suit, Nomex, Flame-Resistant, Size 40 Long, 1 ea

Catalog No. 5000792954
Encompass

This flight suit is designed for demanding environments, offering flame-resistant and flame-retardant properties. It features a durable zipper closure and multiple pockets for functionality, adhering to military specifications.

  • Garment style: flight suit
  • Garment type: flame-resistant, flame-retardant
  • Size: 40 long
  • Color: green
  • Material: Nomex
  • Hazardous protection level: non-hazardous protection
  • Cuff style: open
  • Ankle style: open
  • Seam style: flat felled
  • Adjustable waist belt with hook & loop closure
  • Knife pocket on left leg inseam
  • Bi-swing back
  • Sewn to military specification FNS/PD 96-17 (MIL-C-83141A)
  • Chest size: 40 inch
  • Closure type: zipper
  • Number of pockets: 6

Catalog No. 50-007-92954 Supplier MSC Supplier No. 54612973
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This flight suit is designed for demanding environments, offering flame-resistant and flame-retardant properties. It features a durable zipper closure and multiple pockets for functionality, adhering to military specifications.

  • Garment style: flight suit
  • Garment type: flame-resistant, flame-retardant
  • Size: 40 long
  • Color: green
  • Material: Nomex
  • Hazardous protection level: non-hazardous protection
  • Cuff style: open
  • Ankle style: open
  • Seam style: flat felled
  • Adjustable waist belt with hook & loop closure
  • Knife pocket on left leg inseam
  • Bi-swing back
  • Sewn to military specification FNS/PD 96-17 (MIL-C-83141A)
  • Chest size: 40 inch
  • Closure type: zipper
  • Number of pockets: 6

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.